Learn More
Ponceau S, 0.1% v/v soln. in 5% acetic acid
Description
Ponceau S is used to prepare a stain for rapid reversible detection of protein bands on nitrocellulose or polyvinylidene fluoride membranes, as well as on cellulose acetate membranes. And it is also used as a rapid protein band stain.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Red |
| Concentration | 0.2% v/v soln. in 5% acetic acid |
| Quantity | 500 mL |
| Physical Form | Liquid |
| Solubility Information | Soluble in water (1mg/ml). Insoluble in ether. |
| Formula Weight | 760.56 |
| Chemical Name or Material | Ponceau S |
Safety and Handling
- Ponceau S, 0.1% v/v soln. in 5% acetic acid
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contain: Acetic acid, PonceauS
RUO – Research Use Only
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.