Learn More
Ponceau S, 0.1% v/v soln. in 5% acetic acid
Description
Ponceau S is used to prepare a stain for rapid reversible detection of protein bands on nitrocellulose or polyvinylidene fluoride membranes, as well as on cellulose acetate membranes. And it is also used as a rapid protein band stain.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Red |
| Concentration | 0.2% v/v soln. in 5% acetic acid |
| Solubility Information | Soluble in water (1mg/ml). Insoluble in ether. |
| Formula Weight | 760.56 |
| Quantity | 1 L |
| Physical Form | Liquid |
| Chemical Name or Material | Ponceau S |
Safety and Handling
- Ponceau S, 0.1% v/v soln. in 5% acetic acid
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contain: Acetic acid, PonceauS
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.