missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Porcine IL-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB652
This item is not returnable.
View return policy
Description
IL-2 Monoclonal specifically detects IL-2 in Porcine samples. It is validated for Western Blot.
Specifications
| IL-2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| aldesleukin, IL2, IL-2, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, Proleukin, T cell growth factor, T-cell growth factor, TCGF | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 3558 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot | |
| 100321 | |
| Western Blot 1 ug/mL | |
| P26891 | |
| IL2 | |
| E. coli-derived recombinant porcine IL-2 Ala21-Thr154 Accession # P26891 | |
| 500 μg | |
| Primary | |
| Detects porcine IL-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human IL-2, recombinant mouse IL-2, and recombinant rat IL-2 is observed and no cross-reactivity with recombinant cotton rat IL-2 is observed. | |
| Pig | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction