missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium 4-bromophenyltrifluoroborate, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Potassium 4-bromophenyltrifluoroborate is used as potent boronic acid surrogates in Suzuki-Miyaura Cross-Coupling reactions.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under inert gas. Moisture sensitive. Incompatible with strong oxidizing agents.
Potassium 4-bromophenyltrifluoroborate is used as potent boronic acid surrogates in Suzuki-Miyaura Cross-Coupling reactions.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under inert gas. Moisture sensitive. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | >300°C |
| Quantity | 1 g |
| Beilstein | 9297533 |
| Solubility Information | Soluble in water. |
| IUPAC Name | potassium (4-bromophenyl)trifluoroboranuide |
| Formula Weight | 262.91 |
| Percent Purity | 97% |
| Chemical Name or Material | Potassium 4-bromophenyltrifluoroborate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction