missing translation for 'onlineSavingsMsg'
Learn More

Potassium 4-iodophenyltrifluoroborate, 96%

Product Code. 11492130
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11492130

Brand: Thermo Scientific Alfa Aesar H28758.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 912350-00-6 | C6H4BF3IK | 309.906 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 912350-00-6
Molecular Formula C6H4BF3IK
Molecular Weight (g/mol) 309.906
MDL Number MFCD09800738
InChI Key WTXNSBOVCAXUGV-UHFFFAOYSA-N
Synonym potassium 4-iodophenyltrifluoroborate, potassium trifluoro 4-iodophenyl boranuide, amtb080, potassium trifluoro 4-iodophenyl borate 1-
PubChem CID 23688160
IUPAC Name potassium;trifluoro-(4-iodophenyl)boranuide
SMILES [B-](C1=CC=C(C=C1)I)(F)(F)F.[K+]

Specifications

Melting Point 297°C to 299°C
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 309.9
Percent Purity 96%
Chemical Name or Material Potassium 4-iodophenyltrifluoroborate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.