missing translation for 'onlineSavingsMsg'
Learn More

Potassium 4-(trifluoromethyl)phenyltrifluoroborate, 96%, Thermo Scientific™

Product Code. 15430526
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15430526

Brand: Thermo Scientific Alfa Aesar H53259.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Reactant in the Suzuki-Miyaura Cross-Coupling of Aryl Imidazolylsulfonates in water. Reactant in cross-coupling reaction of potassium aryltrifluoroborates with organic chlorides in aqueous media catalyzed by an oxime-derived palladacycle.

Solubility
High air and moisture stability. Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 166328-08-1
Molecular Formula C7H4BF6K
Molecular Weight (g/mol) 252.008
MDL Number MFCD04039955
InChI Key KVDSXGFQLXKOID-UHFFFAOYSA-N
Synonym potassium 4-trifluoromethyl phenyltrifluoroborate, potassium trifluoro 4-trifluoromethyl phenyl borate, potassium trifluoro 4-trifluoromethyl phenyl boranuide, pubchem11514, potassium 4-trifluoromethylphenyl trifluoroborate, potassium 4-trifluoromethylphenyl trifluoroborate,, potassium 4-trifluoromethyl phenyl trifluoroborate, potassium trifluoro-4-trifluoromethyl phenyl boranuide, potassium trifluoro 4-trifluoromethyl phenyl borate 1-, potassium ion trifluoro 4-trifluoromethyl phenyl boranuide
PubChem CID 23679121
IUPAC Name potassium;trifluoro-[4-(trifluoromethyl)phenyl]boranuide
SMILES [B-](C1=CC=C(C=C1)C(F)(F)F)(F)(F)F.[K+]

Specifications

Melting Point 280°C to 286°C
Quantity 5 g
Solubility Information High air and moisture stability. Slightly soluble in water.
Formula Weight 252.01
Percent Purity 96%
Chemical Name or Material Potassium 4-(trifluoromethyl)phenyltrifluoroborate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.