missing translation for 'onlineSavingsMsg'
Learn More

Potassium bromate, 99+%, extra pure

Product Code. 10474335 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
2.5kg
500g
Product Code. Quantity Packaging unitSize
10474335 2.5 kg Plastic bottle 2.5kg
10338490 500 g Plastic bottle 500g
2 options
This item is not returnable. View return policy

Product Code. 10474335

Brand: Thermo Scientific Acros 208850025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7758-01-2 | BrKO3 | 167.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent, Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1-2-7758
Molecular Formula BrKO3
Molecular Weight (g/mol) 167.01
MDL Number MFCD00011359
InChI Key OCATYIAKPYKMPG-UHFFFAOYSA-M
Synonym potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat
PubChem CID 23673461
ChEBI CHEBI:38211
IUPAC Name potassium;bromate
SMILES [O-]Br(=O)=O.[K+]

Specifications

Melting Point 350.0°C
Color Colorless
Physical Form Liquid
Quantity 2.5 kg
Assay Percent Range 99% min. (Iodometry)
Linear Formula KBrO3
Fieser 11,431
Merck Index 15, 7738
Solubility Information Solubility in water: 70g/L (20°C). Other solubilities: practically insoluble in alcohol,soluble in boiling water
Formula Weight 167.01
Percent Purity ≥99%
Grade Extra Pure
Packaging Plastic bottle
Specific Gravity 3.27
Density 3.2700g/mL
Chemical Name or Material Potassium bromate, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.