missing translation for 'onlineSavingsMsg'
Learn More

Potassium bromate, 99%

Product Code. 11403807
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11403807 500 g 500g
11493797 100 g 100g
11413807 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11403807 Supplier Thermo Scientific Alfa Aesar Supplier No. A18258.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7758-01-2 | BrKO3 | 167.00 g/mol

Used as an analytical reagent and an oxidizing agent. Potassium bromate is used primarily as a maturing agent for flour and as a dough conditioner. It is also used as a laboratory reagent, in permanent-wave compounds such as food additives and explosives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as an analytical reagent and an oxidizing agent. Potassium bromate is used primarily as a maturing agent for flour and as a dough conditioner. It is also used as a laboratory reagent, in permanent-wave compounds such as food additives and explosives.

Solubility
Slightly soluble in alcohol but insoluble in acetone and ethanol.

Notes
Strong oxidizer. Incompatible with combustible materials, organics, reducing agents, aluminum and finely powdered metals
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1-2-7758
Molecular Formula BrKO3
Molecular Weight (g/mol) 167.00
MDL Number MFCD00011359
InChI Key OCATYIAKPYKMPG-UHFFFAOYSA-M
Synonym potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat
PubChem CID 23673461
ChEBI CHEBI:38211
IUPAC Name potassium;bromate
SMILES [O-]Br(=O)=O.[K+]

Specifications

Melting Point ∼370°C (decomposition)
Quantity 500 g
UN Number UN1484
Merck Index 14,7617
Solubility Information Slightly soluble in alcohol but insoluble in acetone and ethanol.
Formula Weight 167
Percent Purity 99%
Odor Odorless
Density 3.27
Chemical Name or Material Potassium bromate
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Potassium bromate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 1B
  • Oxidising solid Category 1
Hazard Statement
  • H271-May cause fire or explosion; strong oxidiser.
  • H301-Toxic if swallowed.
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Potassium bromate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.