Learn More
Potassium bromate, Acculute Standard Volumetric Solution, Final Concentration 0.1N, Thermo Scientific Chemicals
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Specifica
| Color | Colorless |
| CAS | 7758-01-2, 7732-18-5 |
| Quantity | 1 U |
| UN Number | UN3139 |
| Physical Form | Liquid |
| Formula Weight | 167 |
| Chemical Name or Material | Potassium bromate Acculute Standard Volumetric Solution, Final Concentration 0.1N |
Frequently Asked Questions (FAQs)
The following is the dilution protocol:
- Transfer entire contents of one unit bottle into a 1L volumetric flask.
- Rinse bottle residue, the threads of the bottle, and inner bottle cap each with DI water, and transfer to the 1L flask. Repeat rinse three times.
- Bring the 1L flask to volume with DI water and mix well.
The final solution in the 1L flask has a concentration of 0.1N Potassium bromate.
1 unit of Potassium bromate, Acculute Standard Volumetric Solution, Final Concentration 0.1N, when diluted to 1 L, will yield a final concentration of 0.1N Potassium bromate solution.
RUO – Research Use Only
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.