Learn More
Potassium bromate, ACS, 99.8% min
Description
Used as an analytical reagent and an oxidizing agent. Potassium bromate is used primarily as a maturing agent for flour and as a dough conditioner. It is also used as a laboratory reagent, in permanent-wave compounds such as food additives and explosives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 370°C (decomposition) |
| Physical Form | Granules |
| Quantity | 100 g |
| Assay Percent Range | 99.8% min |
| Solubility Information | Slightly soluble in alcohol but insoluble in acetone and ethanol. |
| Formula Weight | 167 |
| Grade | ACS Reagent |
| Density | 3.27 g/mL |
| Chemical Name or Material | Potassium bromate |
Safety and Handling
- Potassium bromate
- Danger
- Acute toxicity Category 3
- Carcinogenicity Category 1B
- Oxidising solid Category 1
- H271-May cause fire or explosion; strong oxidiser.
- H301-Toxic if swallowed.
- H350-May cause cancer.
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Potassium bromate
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.