missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium bromate/Potassium bromide, 0.1N Standardized Solution
Click to view available options
Quantity:
1 L
4 L
Conditionnement:
1 L
4 L
Description
Potassium bromate/Potassium bromide is used as an analytical reagent for volumetric analysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécification
Spécification
| Odor | Odorless |
| Physical Form | Liquid |
| Color | Colorless |
| Solubility | Miscible with water |
| Quantity | 1 L |
| Chemical Name or Material | Potassium bromate/Potassium bromide |
Sécurité et traitement
Identifiant produit
- Potassium bromate/Potassium bromide, 0.1N Standardized Solution
- Danger
- Carcinogenicity Category 1B
- H350-May cause cancer.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Potassium bromide, Potassium bromate
RUO – Research Use Only
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu