missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bromate solution 0.016mol/L, Fisher Chemical™

Product Code. 12903574 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
Product Code. Quantity unitSize
12903574 1000 mL 1000mL
1 options
This item is not returnable. View return policy

Product Code. 12903574

Brand: Fisher Chemical VL 253 1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Bottle
CAS 1-2-7758
Molecular Formula BrKO3
Concentration 0.016 to 0.017 M
InChI Key OCATYIAKPYKMPG-UHFFFAOYSA-M
SMILES [O-]Br(=O)=O.[K+]
IUPAC Name potassium;bromate
Molecular Weight (g/mol) 167.00
PubChem CID 23673461
Quantity 1000 mL
ChEBI CHEBI:38211
Chemical Name or Material Potassium Bromate
Show More Show Less
compliance-icons
Product Identifier
  • Potassium bromate 0.1N (1/60M). Volumetric Solution
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
Hazard Statement
  • H350-May cause cancer.
Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.