missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bromide solution 0.5mol/L, Fisher Chemical™

Product Code. 12913574 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12913574 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12913574 Supplier Fisher Chemical Supplier No. VL 254 1000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Bottle
CAS 7758-02-3, 7732-18-5
Molecular Formula BrK
Concentration 0.5 M
MDL Number MFCD00011358
Synonym potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
SMILES [K+].[Br-]
IUPAC Name potassium bromide
Molecular Weight (g/mol) 119.00
PubChem CID 253877
Quantity 1000 mL
ChEBI CHEBI:32030
Chemical Name or Material Potassium Bromide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.