missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse APOA1BP ELISA Kit (Colorimetric)

Product Code. 18605491 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18605491 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18605491 Supplier R&D Systems™ Supplier No. NBP2770341Kit
  • ONLINE EXCLUSIVE PRICE
    1135.65 €
    991.20 € / 1 sets
    Save 144.45 €
    12% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Kit (Colorimetric)

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 9.375 pg/mL to 600 pg/mL
Assay Sensitivity 2.34 pg/mL
Product Line Novus
Product Type ELISA Kit (Colorimetric)
Sample Type Serum, Plasma, Tissue Homogenates
Specificity This assay has high sensitivity and excellent specificity for detection of Mouse Apolipoprotein A-I/ApoA1. No significant cross-reactivity or interference between Mouse Apolipoprotein A-I/ApoA1 and analogues was observed.
For Use With (Application) ELISA
Gene ID (Entrez) 128240
Gene Symbol NAXE
Quantity 1 Kit
Research Discipline Cardiovascular Biology
Storage Requirements Store at 4°C.
Synonym AIBPAI-BP, apolipoprotein A-I binding protein, apolipoprotein A-I-binding protein, MGC119143, MGC119144, MGC119145, YjeF N-terminal domain-containing protein 1, YjeF_N1, YJEFN1apoA-I binding protein
Target Mouse
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.