missing translation for 'onlineSavingsMsg'
Learn More

Potassium Dichromate solution, 0.0167M EU Pharmacopoeia, Fisher Chemical™
BIOPHARMA

Product Code. 12950874 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12950874 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 12950874 Supplier Fisher Chemical Supplier No. VL 037 1000
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Specifications

Packaging Bottle
CAS 7732-18-5
Concentration 0.0167 M
Quantity 1000 mL
Chemical Name or Material Potassium dichromate solution
compliance-icons compliance-icons
Product Identifier
  • Potassium dichromate 0.0167M. Volumetric Solution European Pharmacopoeia Ref: 3004600
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Germ cell mutagenicity Category 1B
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 1B
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H332-Harmful if inhaled.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H360-May damage fertility or the unborn child.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.