missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ ADAR Recombinant Protein Antigen

Product Code. 18605547 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18605547 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18605547 Supplier R&D Systems™ Supplier No. NBP305523PEP
  • ONLINE EXCLUSIVE PRICE
    313.07 €
    246.75 € / 100µL
    Save 66.32 €
    21% Off
 Restricted Item
Estimated Shipment: 26-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Antibody Competition
Formulation PBS and 1 M Urea, pH 7.4.
Gene ID (Entrez) 103
Molecular Weight (g/mol) MolecularWeight-theroretical: 34 kDa
Quantity 100 μL
Storage Requirements Store at -20°C. Avoid freeze/thaw cycles.
Gene Alias ADAR1DSH, adenosine deaminase, RNA-specific, DRADAP136, DSRADadenosine deaminase acting on RNA 1-A, EC 3.5.4, EC 3.5.4.-, G1P1double-stranded RNA-specific adenosine deaminase, IFI-4, IFI4dsRNA adenosine deaminase, interferon-induced protein 4, Interferon-inducible protein 4, K88DSRBP, p136,136 kDa double-stranded RNA-binding protein
Gene Symbol ADAR
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Inhibitor Properties This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP3-05523.It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/mLFor current lot information, including availability, please contact our technical support team click nb-technical@bio-techne.com For further blocking tide related information and a protocol, click here .
Form Purified
Product Line Novus
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein ADAR
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.