missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydrogensulfate, Fused, ACS Reagent, Mixture of K2S2O7 and KHSO4, Honeywell Fluka™

Product Code. 15672230 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly Bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15672230 250 g Poly Bottle 250g
15662230 1 kg Poly Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15672230 Supplier Honeywell-Fluka Supplier No. 223697250G
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Reagent, Mixture of K2S2O7 and KHSO4

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7646-93-7 , 7790-62-7
Molecular Formula HKO4S
Molecular Weight (g/mol) 136.16
InChI Key CHKVPAROMQMJNQ-UHFFFAOYSA-M
IUPAC Name potassium hydrogen sulfate
SMILES [K+].OS([O-])(=O)=O

Specifications

Color White
Quantity 250 g
MDL Number MFCD00011404
UN Number UN2509
Grade ACS Reagent
Packaging Poly Bottle
Appearance Powder
Density 2.512g/mL at 25°C
Chemical Name or Material Potassium Hydrogensulfate, Fused, Mixture of K2S2O7 and KHSO4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.