Learn More
Potassium iodate, 0.1N Standardized Solution
Description
Potassium iodate 0.1N solution is used as a standard solution in volumetric titrations. It is extensively used in analytical chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Colorless |
| CAS | 5-6-7758 |
| Concentration | 0.1N |
| Solubility Information | Miscible with water,potassium iodide solution and sulfuric acid. |
| Formula Weight | 214 |
| Odor | Odorless |
| Quantity | 4 L |
| Physical Form | Liquid |
| Chemical Name or Material | Potassium iodate, Standardized Solution |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.