missing translation for 'onlineSavingsMsg'
Learn More

Potassium L-tartrate hemihydrate, 99%

Product Code. 11431948
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11431948 5000 g 5000g
11421948 250 g 250g
11411948 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11431948 Supplier Thermo Scientific Alfa Aesar Supplier No. A14556.0I
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6100-19-2 | C8H14K4O13 | 474.58 g/mol

Potassium L-tartrate hemihydrate is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium L-tartrate hemihydrate is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6100-19-2
Molecular Formula C8H14K4O13
Molecular Weight (g/mol) 474.58
MDL Number MFCD00150136,MFCD00150136
InChI Key DFVWEDQSAWCYPI-UHFFFAOYNA-N
Synonym potassium tartrate dibasic hemihydrate
PubChem CID 131855961
SMILES O.[K].[K].[K].[K].OC(C(O)C(O)=O)C(O)=O.OC(C(O)C(O)=O)C(O)=O

Specifications

Melting Point 155°C (in -H2O)
Density 1.98
Boiling Point 200°C to 220°C
Quantity 5000 g
Beilstein 6119985
Merck Index 14,7677
Solubility Information Soluble in water.
IUPAC Name bis(2,3-dihydroxybutanedioic acid) hydrate tetrapotassium
Formula Weight 235.29 (226.27 Anhydrous)
Percent Purity 99%
Chemical Name or Material Potassium L-tartrate hemihydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.