missing translation for 'onlineSavingsMsg'
Learn More

Potassium oxonate, MedChemExpress

Product Code. 16479847
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16479847 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16479847 Supplier Medchem Express Supplier No. HY17511
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Potassium oxonate is an inhibitor of uricase, inhibits the phosphorylation of 5-FU to 5-fluorouridine-5'-monophosphate catalyzed by pyrimidine phosphoribosyl-transferase in a different manner from allopurinol in cell-free extracts and intact cells in vitro.

Potassium oxonate is an inhibitor of uricase, inhibits the phosphorylation of 5-FU to 5-fluorouridine-5'-monophosphate catalyzed by pyrimidine phosphoribosyl-transferase in a different manner from allopurinol in cell-free extracts and intact cells in vitro.

Specifications

Chemical Name or Material Potassium oxonate
Physical Form Solid
CAS 2207-75-2
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C4H2KN3O4
For Use With (Application) Metabolism-protein/nucleotide metabolism
Synonym Potassium azaorotate Potassium otastat
Solubility Information H2O : 5 mg/mL (25.62 mM; ultrasonic and warming and heat to 80°C) ∣DMSO : < 1 mg/mL (ultrasonic;warming;heat to 80°C) (insoluble or slightly soluble)
SMILES O=C(O[K])C1=NC(O)=NC(O)=N1
Molecular Weight (g/mol) 195.17
Formula Weight 195.17
Percent Purity 97.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.