missing translation for 'onlineSavingsMsg'
Learn More

Potassium permanganate, ACS, 99.0% min, Thermo Scientific™

For Use in Photography, Tanning Leather, Purifying Water

Brand:  Thermo Scientific Alfa Aesar 014307.36

Quantity 500g

 View more versions of this product

Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.

Product Code. 11307877

  • 69.70 € / 500g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

7722-64-7
158.032
VZJVWSHVAAUDKD-UHFFFAOYSA-N
516875
[O-][Mn](=O)(=O)=O.[K+]
KMnO4
MFCD00011364
potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo
potassium;permanganate

Specifications

7722-64-7
99.0% min.
MFCD00011364
Partially soluble in water. Decomposes in organic solvents and acids.
[O-][Mn](=O)(=O)=O.[K+]
158.032
158.03
Light sensitive
Decomposes in organic solvents and acids.
Potassium permanganate
500g
KMnO4
potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo
VZJVWSHVAAUDKD-UHFFFAOYSA-N
potassium;permanganate
516875
ACS
Dark crystalline
2.7 g/cm3
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.