Learn More
Potassium permanganate, ACS, 99.0% min, Thermo Scientific™
For Use in Photography, Tanning Leather, Purifying Water
25.80 € - 190.00 €
Chemical Identifiers
| CAS | 7722-64-7 |
|---|---|
| Molecular Formula | KMnO4 |
| Molecular Weight (g/mol) | 158.032 |
| MDL Number | MFCD00011364 |
| InChI Key | VZJVWSHVAAUDKD-UHFFFAOYSA-N |
| Synonym | potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo |
| PubChem CID | 516875 |
| IUPAC Name | potassium;permanganate |
| SMILES | [O-][Mn](=O)(=O)=O.[K+] |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
11317877
|
Thermo Scientific Alfa Aesar
014307.22 |
100g |
25.80 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
11307877
|
Thermo Scientific Alfa Aesar
014307.36 |
500g |
69.70 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
11397867
|
Thermo Scientific Alfa Aesar
014307.A3 |
2kg |
190.00 €
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| KMnO4 | |
| MFCD00011364 | |
| potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo | |
| potassium;permanganate |
Specifications
| 7722-64-7 | |
| KMnO4 | |
| potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo | |
| VZJVWSHVAAUDKD-UHFFFAOYSA-N | |
| potassium;permanganate | |
| 516875 | |
| ACS | |
| Dark crystalline | |
| 2.7 g/cm3 |
| 99.0% min. | |
| MFCD00011364 | |
| Partially soluble in water. Decomposes in organic solvents and acids. | |
| [O-][Mn](=O)(=O)=O.[K+] | |
| 158.032 | |
| 158.03 | |
| Light sensitive | |
| Decomposes in organic solvents and acids. | |
| Potassium permanganate |
Safety and Handling
This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
P201-P202-P210-P220-P221-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H272-H302-H314-H335-H361d-H373
missing translation for 'dotInformation' : Hazard Class: 5.1, Packing Group: II
missing translation for 'einecsNumber' : 231-760-3
missing translation for 'rtecsNumber' : SD6475000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Potassium permanganate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Oxidising solid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 1 C
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H272-May intensify fire; oxidiser.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H361d-Suspected of damaging the unborn child.
- H373-May cause damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contain: Potassium permanganate
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.