missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phenolate, TRC

Product Code. 30418853
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30418853 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30418853 Supplier Toronto Research Chemicals Supplier No. TRCP9929331G
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-67-4
Molecular Formula C6H5KO
Molecular Weight (g/mol) 132.2
Synonym Phenoxypotassium, Potassium Carbolate, Potassium Phenate, Potassium Phenylate
IUPAC Name potassium;phenoxide
SMILES [K+].[O-]c1ccccc1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6H6O.K/c7-6-4-2-1-3-5-6;/h1-5,7H;/q;+1/p-1
Formula Weight 131.9977
Chemical Name or Material Potassium Phenolate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.