missing translation for 'onlineSavingsMsg'
Learn More

Potassium Standard: K @ 1000 μg/mL in 5% HNO3, VHG™

Product Code. 17968889
Change view
Click to view available options
Quantity:
50 mL
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
50mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17968889 50 mL 50mL
17967079 250 mL 250mL
17968519 500 mL 500mL
17978879 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 17968889 Supplier VHG Labs Supplier No. VHGPKN50
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Potassium Standard: K @ 1000 μg/mL in 5% HNO3
TRUSTED_SUSTAINABILITY

Spécification

Chemical Name or Material Potassium
CAS 9-7-7440
Methods ICP/ICP-MS
Analyte or Component Names Potassium
Shipping Condition Room Temperature
Molecular Formula K
Certifications/Compliance ISO 17034
Synonym Kalium
InChI Formula InChI=1S/K.H
SMILES [K]
IUPAC Name potassium
Solvent or Matrix Dilute HNO3
Molecular Weight (g/mol) 39.1
Quantity 50 mL
Formula Weight 38.9637 g/mol
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.