missing translation for 'onlineSavingsMsg'
Learn More

Potassium tetrakis(pentafluorophenyl)borate, 97%

Product Code. 15417667
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15417667 1 g 1g
15427667 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15417667 Supplier Thermo Scientific Alfa Aesar Supplier No. H36527.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 89171-23-3 | C24BF20K | 718.14 g/mol

Potassium tetrakis(pentafluorophenyl)borate is used as a biochemical reagent, a catalyst and a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium tetrakis(pentafluorophenyl)borate is used as a biochemical reagent, a catalyst and a pharmaceutical intermediate.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89171-23-3
Molecular Formula C24BF20K
Molecular Weight (g/mol) 718.14
MDL Number MFCD06797410
InChI Key GYBHRIJOPWTIKA-UHFFFAOYSA-N
Synonym potassium tetrakis pentafluorophenyl borate, potassium tetrakis pentafluorophe-nyl borate, tetrakis-pentafluorophenyl-potassium borate, potassium tetrakis 2,3,4,5,6-pentafluorophenyl boranuide
PubChem CID 23693577
IUPAC Name potassium;tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide
SMILES [B-](C1=C(C(=C(C(=C1F)F)F)F)F)(C2=C(C(=C(C(=C2F)F)F)F)F)(C3=C(C(=C(C(=C3F)F)F)F)F)C4=C(C(=C(C(=C4F)F)F)F)F.[K+]

Specifications

Melting Point >300°C
Quantity 1 g
Formula Weight 718.13
Percent Purity 97%
Chemical Name or Material Potassium tetrakis(pentafluorophenyl)borate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.