missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POU3F2/OCT7 Antibody (CL6228) [Janelia Fluor™ 646], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61436JF646
This item is not returnable.
View return policy
Description
POU3F2/OCT7 Monoclonal antibody specifically detects POU3F2/OCT7 in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown,Immunocytochemistry/ Immunofluorescence
Specifications
| POU3F2/OCT7 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| Brain-2, Brain-specific homeobox/POU domain protein 2, brn-2, BRN2brain-2, Nervous system-specific octamer-binding transcription factor N-Oct-3, Oct-7, OCT7POU domain class 3, transcription factor 2, Octamer-binding protein 7, Octamer-binding transcription factor 7, OTF7, OTF-7, POU class 3 homeobox 2, POU domain, class 3, transcription factor 2, POUF3 | |
| This antibody was developed using a recombinant protein derived from P20265, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Transcription Factors and Regulators | |
| 5454 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown, Immunocytochemistry/Immunofluorescence | |
| CL6228 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction