missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POU3F3 Antibody (CL15308) [Unconjugated], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43660-25ul
This item is not returnable.
View return policy
Description
POU3F3 Monoclonal antibody specifically detects POU3F3 in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| POU3F3 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, Immunohistochemistry 1:1000-1:2500, Immunocytochemistry/ Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:1000-1:2500 | |
| brain-1, Brain-specific homeobox/POU domain protein 1, Brn-1, BRN1brn-1, Oct-8, Octamer-binding protein 8, Octamer-binding transcription factor 8, OTF-8, OTF8oct-8, POU class 3 homeobox 3, POU domain class 3, transcription factor 3, POU domain, class 3, transcription factor 3 | |
| This antibody was generated using a synthetic peptide of P20264, with the exact immunogen sequence remaining proprietary. | |
| 25 μL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL15308 | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 5455 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction