missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POU4F1 Recombinant Rabbit Monoclonal Antibody (9A5O9)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557983
This item is not returnable.
View return policy
Description
Immunogen sequence: MMSMNSKQPH FAMHPTLPEH KYPSLHSSSE AIRRACLPTP PLQSNLFASL DETLLARAEA LAAVDIAVSQ GKSHPFKPDA TYHTMNSVPC TSTSTVPLAH.
BRN3A is a class IV POU domain-containing transcription factor highly expressed in the developing sensory nervous system and in cells of the B- and T-lymphocytic lineages.
Specifications
| POU4F1 | |
| Recombinant Monoclonal | |
| 1.04 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Q01851 | |
| Pou4f1 | |
| Synthetic Peptide. | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 9A5O9 | |
| Unconjugated | |
| Pou4f1 | |
| Brain-3A; brain-specific homeobox/POU domain protein 3A; Brn3; Brn-3; brn3.0; Brn-3.0; Brn3a; brn-3A; brn3b; E130119J07Rik; etID22526.1; FLJ13449; homeobox/POU domain protein RDC-1; LOW QUALITY PROTEIN: POU domain, class 4, transcription factor 1; Oct-T1; POU class 4 homeobox 1; POU domain, class 4, transcription factor 1; POU4F1; pou4f1 protein; RDC1; RDC-1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 5457 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction