Learn More
Pour Point Standard IP15, ASTM D97, ISO 3016, BS 2000, Pt 15 (nominal value: -30.3 °C) UKAS ISO 17034 Certified 250mL, Paragon Scientific™
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
Description
- Certified according to ISO 17025 / ISO 17034 under UKAS accreditation by means of a method specific inter-laboratory study.
- Fully traceable to international standards.
- Supplied with MSDS.
- Tamper evident security packaging.
- 12 month shelf life.
- Manufactured in the United Kingdom
Certifications
Nominal values quoted
Specifications
Specifications
| Quantity | 250 mL |
Nominal values quoted. Certified Values will vary from batch to batch.