missing translation for 'onlineSavingsMsg'
Learn More

Povidone iodine, MedChemExpress

Product Code. 16422399
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16422399 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16422399 Supplier Medchem Express Supplier No. HYB2234
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Povidone iodine (iodopovidone) displays excellent antibacterial activity which can against MRSA and MSSA strains with MICs of 31.25 mg/L and 7.82 mg/L, respectively.

Povidone iodine (iodopovidone) displays excellent antibacterial activity which can against MRSA and MSSA strains with MICs of 31.25 mg/L and 7.82 mg/L, respectively.

Specifications

Chemical Name or Material Povidone iodine
Physical Form Solid
CAS 25655-41-8
Color Red
Quantity 1 g
Purity Grade Notes Research
For Use With (Application) Neuroscience-Neuromodulation
Synonym iodopovidone
Solubility Information DMSO : 15 mg/mL (Need ultrasonic) ∣H2O : 2.4 mg/mL (Need ultrasonic)
SMILES O=C1N(C(C)CC)CCC1.I[F,Cl,Br,I]I.[n].[n:x].[=].[10:1]
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.