missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PowderTainer™ II 1 to 25 kg Single-Use BPC for powder applications, 1 kg

Product Code. 30040289 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
1 kg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
30040289 1 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30040289 Supplier Thermo Scientific™ Supplier No. SH30737.01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The 1 kg Thermo Scientific™ Powdertainer™ II BioProcess Container (BPC) is a 2D powder handling system composed of CX3-9 film that includes 2 ports (1 wash down line, 1 powder port).

The 1 kg Thermo Scientific™ Powdertainer™ II BioProcess Container (BPC) is a 2D powder handling system composed of CX3-9 film that includes 2 ports (1 wash down line, 1 powder port). The Powdertainer BPC is specifically designed for powder containment and discharge applications, and maintains a closed system for maximum recovery of powder while minimizing the risk of cross contamination.

Features:

  • Three-inch tri-clamp port designed for secure connection to, and easy integration, with existing formulation vessels.
  • Powdertainer BPC features suspensor handle for support during discharge, as well as neck clamp to retain powder prior to discharge.
  • Use this system to eliminate post-use cleaning steps required with re-useable containers.

Applications of the Thermo Scientific Powdertainer BPC include:

  • Storage and delivery of powder culture media and buffers.
  • Storage and dispensing of pre-weighed chemicals and other process powders.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Powder Application
Includes 2 Ports (1 Wash Down Line, 1 Powder Port)
Tubing Style Silicone
Film Description CX3-9
Port Detail Powder port: 7.6 cm (3 in.) tri-clamp with integrated seal
Product Line PowderTainer™ II
Dimensions (L x W) 16.8 x 12.5 in. (42.7 x 31.8 cm)
Product Type Bioprocess Container for Powder
Capacity (Metric) 1 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.