missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PowderTainer™ II 1 to 25kg Single-Use BPC for powder applications, 25 kg

Product Code. 31932210 Shop All Thermo Scientific Products
Change view
Click to view available options
Port Detail:
Powder Port: 3 in.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Port Detail Unit Size
31932210 Powder Port: 3 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31932210 Supplier Thermo Scientific™ Supplier No. SH30864.03
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

2D powder handling system composed of CX3-9 film that includes 1 port (1 powder port)

  • The Powdertainer BPC is specifically designed for powder containment and discharge applications, and maintains a closed system for maximum recovery of powder while minimizing the risk of cross contamination.
  • Three-inch tri-clamp port designed for secure connection to, and easy integration, with existing formulation vessels.
  • Powdertainer BPC features suspensor handle for support during discharge, as well as neck clamp to retain powder prior to discharge.
  • Use this system to eliminate post-use cleaning steps required with re-useable containers.
  • Storage and delivery of powder culture media and buffers.
  • Storage and dispensing of pre-weighed chemicals and other process powders.

TRUSTED_SUSTAINABILITY

Specifications

Includes 1 Port (1 Powder Port)
Film Description CX3-9
Outer Packaging Packages are placed in strong cardboard cartons with product and lot identification.
Port Detail Powder Port: 3 in.
Product Line PowderTainer™ II
Protective Inner Packaging Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area.
Sterilization Method Non-Irradiated
Tubing Line 1 Washdown Line: N/A
Tubing Line 2 Powder port: 7.6 cm (3 in.) tri-clamp with intergrated seal
Dimensions (L x W x H) 83.3 x 59.2 cm (32.8 x 23.3 in.)
Width (English) 23.3 in.
Weight (Metric) 25 kg
Product Type Bioprocess Container for Powder
Length (Metric) 83.3 cm
Width (Metric) 59.2 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.