missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ PowderTainer™ II 1 to 25kg Single-Use BPC for powder applications, 5 kg
Shop All Thermo Scientific ProductsDescription
- A 2D powder handling system composed of CX3-9 film that includes 1 port (1 powder port)
- Specifically designed for powder containment and discharge applications
- Maintains a closed system for maximum recovery of powder while minimizing the risk of cross contamination
Features
- 3 in. tri-clamp port designed for secure connection to, and easy integration with existing formulation vessels
- Powdertainer BPC features suspensor handle for support during discharge, as well as neck clamp to retain powder prior to discharge
- Use this system to eliminate post-use cleaning steps required with re-useable containers
Applications
- Storage and delivery of powder culture media and buffers
- Storage and dispensing of pre-weighed chemicals and other process powders
Specifications
Specifications
| Includes | 1 Port (1 Powder Port) |
| Film Description | CX3-9 |
| Outer Packaging | Packages are placed in strong cardboard cartons with product and lot identification. |
| Port Detail | Powder Port: 3 in. |
| Product Line | PowderTainer™ II |
| Protective Inner Packaging | Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area. |
| Sterilization Method | Non-Irradiated |
| Tubing Line 1 | Washdown Line: N/A |
| Tubing Line 2 | Powder port: 7.6 cm (3 in.) tri-clamp with intergrated seal |
| Product Type | Bioprocess Container for Powder |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?