missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PP14/Glycodelin Antibody (SR1372), Novus Biologicals™
Rabbit Monoclonal Antibody
338.00 € - 449.00 €
Specifications
| Antigen | PP14/Glycodelin |
|---|---|
| Clone | SR1372 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18651344
|
Novus Biologicals
NBP3-22049-100ul |
100 μg |
449.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18693315
|
Novus Biologicals
NBP3-22049-50ul |
50 μg |
338.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PP14/Glycodelin Monoclonal antibody specifically detects PP14/Glycodelin in Human samples. It is validated for Western BlotSpecifications
| PP14/Glycodelin | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| alpha uterine protein, GdF, GdS, glycodelin, glycodelin-A, glycodelin-F, glycodelin-S, MGC138509, MGC142288, PEP, Placental protein 14, PP14 protein (placental protein 14), PP14GDPAEGGdA, pregnancy-associated endometrial a, Pregnancy-associated endometrial alpha-2 globulin, pregnancy-associated endometrial alpha-2-globulin, progestagen-associated endometrial protein (placental protein 14, progestagen-associated endometrial proteinPEG, Progesterone-associated endometrial protein | |
| A synthesized peptide derived from human PP14/Glycodelin (Uniprot #: P09466) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1372 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Developmental Biology | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 5047 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title