missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PP5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA519540
This item is not returnable.
View return policy
Description
This antibody is predicted to react with dog and Xenopus laevis based on sequence homology.
This gene encodes a serine/threonine phosphatase which is a member of the protein phosphatase catalytic subunit family. Proteins in this family participate in pathways regulated by reversible phosphorylation at serine and threonine residues; many of these pathways are involved in the regulation of cell growth and differentiation. The product of this gene has been shown to participate in signaling pathways in response to hormones or cellular stress, and elevated levels of this protein may be associated with breast cancer development. Alternative splicing results in multiple transcript variants.
Specifications
| PP5 | |
| Polyclonal | |
| Unconjugated | |
| Ppp5c | |
| ANP receptor; AU020526; EGK_10788; FLJ36922; FLJ55954; OTTHUMP00000165834; PP5; PPP5; Ppp5c; PPT; PP-T; protein phosphatase 5 catalytic subunit; protein phosphatase 5, catalytic subunit; protein phosphatase T; Serine/threonine-protein phosphatase 5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 19060, 5536, 65179 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 1% BSA and 0.02% sodium azide; pH 7.4 | |
| P53041, P53042, Q60676 | |
| Ppp5c | |
| Synthetic peptide conjugated to KLH derived from within residues 350 - 450 of Human PP5. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction