missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPA1 Recombinant Rabbit Monoclonal Antibody (24GB1365)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557431
This item is not returnable.
View return policy
Description
PPA1 Recombinant Monoclonal Antibody for Western Blot, Flow
The protein encoded by this gene is a member of the inorganic pyrophosphatase (PPase) family. PPases catalyze the hydrolysis of pyrophosphate to inorganic phosphate, which is important for the phosphate metabolism of cells. Studies of a similar protein in bovine suggested a cytoplasmic localization of this enzyme.
Specifications
| PPA1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Ppa1 | |
| 2010317E03Rik; cytosolic inorganic pyrophosphatase; diphosphate phosphohydrolase; epididymis secretory sperm binding protein Li 66p; HEL-S-66p; inorganic diphosphatase; inorganic diphosphatase 1; inorganic pyrophosphatase; inorganic pyrophosphatase 1; IOPPP; Pp; PP1; Ppa1; PPase; Pyp; pyrophosphatase (inorganic) 1; pyrophosphatase 1; pyrophosphate phospho-hydrolase; RP11-367H5.1; SID6-8061 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 294504, 5464, 67895 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1365 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q15181, Q9D819 | |
| Ppa1 | |
| A synthesized peptide derived from human Pyrophosphatase 1 (aa 1-45). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction