missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PPA1 Recombinant Rabbit Monoclonal Antibody (JE64-39)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA560186

Product Code. 30485062

  • 441.00 € / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Positive Control: HeLa cell lysate, HepG2 cell lysate, A431 cell lysate, NIH/3T3 cell lysate, RAW264.7 cell lysate, C6 cell lysate. Tissue Specificity: Low tissue specificity. Subcellular Location: Cytoplasm. Sequence Similarities: 97% Mouse/Rat. Predicted band size: 33 kDa.

The protein encoded by this gene is a member of the inorganic pyrophosphatase (PPase) family. PPases catalyze the hydrolysis of pyrophosphate to inorganic phosphate, which is important for the phosphate metabolism of cells. Studies of a similar protein in bovine suggested a cytoplasmic localization of this enzyme.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PPA1
Recombinant Monoclonal
1 mg/mL
TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Q15181, Q9D819
Ppa1
Synthetic peptide within Human Pyrophosphatase 1 aa 1-100.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Western Blot
JE64-39
Unconjugated
Ppa1
2010317E03Rik; cytosolic inorganic pyrophosphatase; diphosphate phosphohydrolase; epididymis secretory sperm binding protein Li 66p; HEL-S-66p; inorganic diphosphatase; inorganic diphosphatase 1; inorganic pyrophosphatase; inorganic pyrophosphatase 1; IOPPP; Pp; PP1; Ppa1; PPase; Pyp; pyrophosphatase (inorganic) 1; pyrophosphatase 1; pyrophosphate phospho-hydrolase; RP11-367H5.1; SID6-8061
Rabbit
Protein A
RUO
294504, 5464, 67895
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.