missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PPA1 Recombinant Rabbit Monoclonal Antibody (K01_1X39)
GREENER_CHOICE

Product Code. p-200113949
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30828435 100 μL
30830457 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30828435 Supplier Invitrogen™ Supplier No. MA563448

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

PPA1 Recombinant Monoclonal Antibody for Western Blot

The protein encoded by this gene is a member of the inorganic pyrophosphatase (PPase) family. PPases catalyze the hydrolysis of pyrophosphate to inorganic phosphate, which is important for the phosphate metabolism of cells. Studies of a similar protein in bovine suggested a cytoplasmic localization of this enzyme.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPA1
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1X39
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 40% glycerol, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene Ppa1
Gene Accession No. Q15181, Q9D819
Gene Alias 2010317E03Rik; cytosolic inorganic pyrophosphatase; diphosphate phosphohydrolase; epididymis secretory sperm binding protein Li 66p; HEL-S-66p; inorganic diphosphatase; inorganic diphosphatase 1; inorganic pyrophosphatase; inorganic pyrophosphatase 1; IOPPP; Pp; PP1; Ppa1; PPase; Pyp; pyrophosphatase (inorganic) 1; pyrophosphatase 1; pyrophosphate phospho-hydrolase; RP11-367H5.1; SID6-8061
Gene Symbols Ppa1
Host Species Rabbit
Immunogen A synthetic peptide of human Pyrophosphatase 1.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5464, 67895
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.