missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPAPDC1B/DPPL1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
415.00 € - 624.00 €
Specifications
| Antigen | PPAPDC1B/DPPL1 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18276362
|
Novus Biologicals
NBP2-55527 |
100 μL |
624.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18648828
|
Novus Biologicals
NBP2-55527-25ul |
25 μL |
415.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PPAPDC1B/DPPL1 Polyclonal specifically detects PPAPDC1B/DPPL1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| PPAPDC1B/DPPL1 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 84513 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:QRLIQPEEMWLYRNPYVEAEYF | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| diacylglycerol pyrophosphate like 1, DPPL1, EC 3.1.3.4, HTPAPphosphatidate phosphatase PPAPDC1B, phosphatidic acid phosphatase type 2 domain containing 1B, Phosphatidic acid phosphatase type 2 domain-containing protein 1B | |
| PLPP5 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title