missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPAR gamma/NR1C3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
302.00 € - 511.00 €
Specifications
| Antigen | PPAR gamma/NR1C3 |
|---|---|
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18040494
|
Novus Biologicals
NBP2-57833 |
100 μL |
511.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18646076
|
Novus Biologicals
NBP2-57833-25ul |
25 μL |
302.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PPAR gamma/NR1C3 Polyclonal specifically detects PPAR gamma/NR1C3 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| PPAR gamma/NR1C3 | |
| Polyclonal | |
| Rabbit | |
| Cancer, mTOR Pathway | |
| CIMT1, NR1C3GLM1, Nuclear receptor subfamily 1 group C member 3, peroxisome proliferator-activated receptor gamma, peroxisome proliferator-activated receptor gamma 1, PPAR gamma, PPARG1peroxisome proliferative activated receptor, gamma, PPARG2PPARgamma, PPAR-gamma | |
| PPARG | |
| IgG | |
| Affinity Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| 5468 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:DYKYDLKLQEYQSAIKVEPASPPYYSEKTQLYNKPHEEPSNSLMAIECRVC | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title