missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPFIA3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94799-0.1ml
This item is not returnable.
View return policy
Description
PPFIA3 Polyclonal antibody specifically detects PPFIA3 in Human samples. It is validated for Western Blot
Specifications
| PPFIA3 | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| KIAA0654liprin, liprin-alpha-3, LPNA3liprin-alpha 3, MGC126567, MGC126569, Protein tyrosine phosphatase receptor type f polypeptide-interacting proteinalpha-3, protein tyrosine phosphatase, receptor type, f polypeptide (PTPRF), interactingprotein (liprin), alpha 3, protein tyrosine phosphatase, receptor type, f polypeptide, alpha 3, PTPRF-interacting protein alpha-3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 460-590 of human PPFIA3 (NP_003651.1). NSLSEEIANMKKLQDELLLNKEQLLAEMERMQMEIDQLRGRPPSSYSRSLPGSALELRYSQAPTLPSGAHLDPYVAGSGRAGKRGRWSGVKEEPSKDWERSAPAGSIPPPFPGELDGSDEEEAEGMFGAEL | |
| 0.1 mL | |
| Protein Phosphatase | |
| 8541 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction