missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPIL1 Antibody (08), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06214-50ul
This item is not returnable.
View return policy
Description
PPIL1 Monoclonal antibody specifically detects PPIL1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunoprecipitation
Specifications
| PPIL1 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 51645 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, Flow Cytometry, ELISA, Immunoprecipitation | |
| 08 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:25-1:100, ELISA 1:1000-1:2000, Immunoprecipitation 4-6 μg/mg of lysate | |
| CYPL1cyclophilin-related gene 1, EC 5.2.1.8, hCyPX, MGC678, peptidyl-prolyl cis-trans isomerase, peptidyl-prolyl cis-trans isomerase-like 1, peptidylprolyl isomerase (cyclophilin)-like 1, PPIase, Rotamase PPIL1 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PPIL1 (Uniprot#: Q9Y3C6; Met1-Gly166). | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction