missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPM1G Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5149035
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
The protein encoded by this gene is a member of the PP2C family of Ser/Thr protein phosphatases. PP2C family members are known to be negative regulators of cell stress response pathways. This phosphatase is found to be responsible for the dephosphorylation of Pre-mRNA splicing factors, which is important for the formation of functional spliceosome. Studies of a similar gene in mice suggested a role of this phosphatase in regulating cell cycle progression. Alternatively spliced transcript variants encoding the same protein have been described.
Specifications
| PPM1G | |
| Polyclonal | |
| Unconjugated | |
| Ppm1g | |
| AI385652; fibroblast growth factor inducible 13; fibroblast growth factor-inducible protein 13; Fin13; PP2C, gamma; PP2CG; PP2CGAMMA; PP2C-gamma; Ppm1c; Ppm1g; PPP2CG; protein phosphatase 1C; Protein phosphatase 1G; protein phosphatase 1G (formerly 2C), magnesium-dependent, gamma isoform; protein phosphatase 2, catalytic subunit, gamma isoform; protein phosphatase 2C gamma isoform; protein phosphatase 2C isoform gamma; Protein phosphatase magnesium-dependent 1 gamma; protein phosphatase, Mg2+/Mn2+ dependent 1G; protein phosphatase, Mg2+/Mn2+ dependent, 1G | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 5496 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| O15355 | |
| Ppm1g | |
| E. coli-derived recombinant human PP2C gamma , Met1-Asp546. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction