missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP2R1A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99603-50ul
This item is not returnable.
View return policy
Description
PPP2R1A Polyclonal antibody specifically detects PPP2R1A in Human samples. It is validated for Western Blot
Specifications
| PPP2R1A | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Medium tumor antigen-associated 61 kDa protein, MGC786, PP2A subunit A isoform PR65-alpha, PP2A subunit A isoform R1-alpha, PP2A-Aalpha, PP2AAALPHA, PR65A, protein phosphatase 2 (formerly 2A), regulatory subunit A (PR 65), alphaisoform, protein phosphatase 2 (formerly 2A), regulatory subunit A, alpha isoform, protein phosphatase 2, regulatory subunit A, alpha, serine/threonine protein phosphatase 2A, 65 kDa regulatory subunit A, alphaisoform, serine/threonine-protein phosphatase 2A 65 kDa regulatory subunit A alphaisoform | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human PPP2R1A. | |
| 50 μg | |
| Protein Phosphatase | |
| 5518 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction