missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP2R1B Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94100-0.1ml
This item is not returnable.
View return policy
Description
PPP2R1B Polyclonal antibody specifically detects PPP2R1B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| PPP2R1B | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:100 | |
| MGC26454, PP2A subunit A isoform PR65-beta, PP2A subunit A isoform R1-beta, PP2A, subunit A, PR65-beta isoform, PP2A, subunit A, R1-beta isoform, PP2A-Abeta, PR65B, protein phosphatase 2 (formerly 2A), regulatory subunit A (PR 65), beta isoform, protein phosphatase 2 (formerly 2A), regulatory subunit A, beta isoform, protein phosphatase 2, regulatory subunit A, beta, protein phosphatase 2, structural/regulatory subunit A, beta, serine/threonine-protein phosphatase 2A 65 kDa regulatory subunit A betaisoform | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 468-667 of human PPP2R1B (NP_859050.1). YAIREAATNNLMKLVQKFGTEWAQNTIVPKVLVMANDPNYLHRMTTLFCINALSEACGQEITTKQMLPIVLKMAGDQVANVRFNVAKSLQKIGPILDTNALQGEVKPVLQKLGQDEDMDVKYFAQEAISVVAQRLRKLEFPVKDSGEPSVPRADKNHFPRPTVPGEDMGKGPVYQLRGDTRDTLAQLGIAELVHFSQSTD | |
| 0.1 mL | |
| Breast Cancer, Protein Phosphatase, Wnt Signaling Pathway | |
| 5519 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction