missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP3CB Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94864-0.1ml
This item is not returnable.
View return policy
Description
PPP3CB Polyclonal antibody specifically detects PPP3CB in Human, Mouse samples. It is validated for Western Blot
Specifications
| PPP3CB | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Calmodulin-dependent calcineurin A subunit beta isoform, CALNA2protein phosphatase 3 (formerly 2B), catalytic subunit, beta isoform(calcineurin A beta), CAM-PRP catalytic subunit, CNA2catalytic subunit, beta isoform, EC 3.1.3.16, protein phosphatase 2B, catalytic subunit, beta isoform, protein phosphatase 3, catalytic subunit, beta isozyme, protein phosphatase from PCR fragment H32, serine/threonine-protein phosphatase 2B catalytic subunit beta isoform | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 286-525 of human PPP3CB (NP_001135825.1). IIRAHEAQDAGYRMYRKSQTTGFPSLITIFSAPNYLDVYNNKAAVLKYENNVMNIRQFNCSPHPYWLPNFMDVFTWSLPFVGEKVTEMLVNVLSICSDDELMTEGEDQFDVGSAAARKEIIRNKIRAIGKMARVFSVLREESESVLTLKGLTPTGMLPSGVLAGGRQTLQSATVEAIEAEKAIRGFSPPHRICSFEEAKGLDRINERMPPRKDAVQQDGFNSLNTAHATENHGTGNHTAQ | |
| 0.1 mL | |
| Cell Cycle and Replication, Wnt Signaling Pathway | |
| 5532 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction