Learn More
PPP5C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11715-1-AP-150UL
 View more versions of this product
Product Code. 16808983
  • 410.00 € / 150µL
Description
This gene encodes a serine/threonine phosphatase which is a member of the protein phosphatase catalytic subunit family. Proteins in this family participate in pathways regulated by reversible phosphorylation at serine and threonine residues; many of these pathways are involved in the regulation of cell growth and differentiation. The product of this gene has been shown to participate in signaling pathways in response to hormones or cellular stress, and elevated levels of this protein may be associated with breast cancer development. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
PPP5C
Polyclonal
Unconjugated
PPP5C
PP T, PP5, PPP5, PPP5C, PPT, Protein phosphatase T
Rabbit
Antigen Affinity Chromatography
RUO
19060, 5536, 65179
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P53041, P53042, Q60676
Ppp5c
PPP5C Fusion Protein Ag2288
150 μL
Primary
Human, Mouse, Rat