missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP6C Antibody (SR1537), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22483-50ul
This item is not returnable.
View return policy
Description
PPP6C Monoclonal antibody specifically detects PPP6C in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence
Specifications
| PPP6C | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunofluorescence | |
| SR1537 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| EC 3.1.3.16, FLJ92648, MGC12249, PP6, PP6C, PPP6, protein phosphatase 6, catalytic subunit, serine/threonine protein phosphatase catalytic subunit, serine/threonine-protein phosphatase 6 catalytic subunit | |
| A synthesized peptide derived from human PPP6C (Uniprot #: O00743) | |
| 50 μg | |
| Cell Cycle and Replication, Signal Transduction | |
| 5537 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction