missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRAME Antibody (CL5146) [Alexa Fluor™ 700], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61148AF700
This item is not returnable.
View return policy
Description
PRAME Monoclonal antibody specifically detects PRAME in Human samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| PRAME | |
| Monoclonal | |
| Alexa Fluor 700 | |
| cancer/testis antigen 130, MAPEOIP4CT130, melanoma antigen preferentially expressed in tumors, OIP-4, Opa-interacting protein 4, Opa-interacting protein OIP4, preferentially expressed antigen in melanoma, Preferentially expressed antigen of melanoma | |
| This antibody was developed using a recombinant protein derived from P78395, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer | |
| 23532 | |
| Store at 4⪚C in the dark. | |
| IgG2a |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| CL5146 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction