Learn More
PRAME Antibody (CL5148) [Janelia Fluor™ 646], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-61149JF646
Product Code. 30291995
  • 737.00 € / 0.10mL
Description
PRAME Monoclonal antibody specifically detects PRAME in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PRAME
Monoclonal
Janelia Fluor 646
cancer/testis antigen 130, MAPEOIP4CT130, melanoma antigen preferentially expressed in tumors, OIP-4, Opa-interacting protein 4, Opa-interacting protein OIP4, preferentially expressed antigen in melanoma, Preferentially expressed antigen of melanoma
This antibody was developed using a recombinant protein derived from P78395, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer
23532
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL5148
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified